4,4-Dimethoxybutan-2-ol 95%
Catalog # APOSOR924280-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Type de conditionnement:Bottle
- Cdt:1 g
- Seq:3047
Specifications
About this item
:
:134.174 g/mol
Storage Temperature:Température ambiante
CAS Number:39562-58-8
Formula:C₆H₁₄O₃
MDL Number:MFCD00046655