ContactUsLinkComponent
Antimicrobial foam soap, Tork
Antimicrobial foam soap, Tork
Catalog # SCAA520800
Supplier:  Essity
Antimicrobial foam soap, Tork
Catalog # SCAA520800
Supplier:  Essity
Specifications
  • Désignation:
    Tork antimicrobial foam soap (Biocide), 1 L
  • Cdt:
    6
Specifications
About this item
Antimicrobial foam soap made of low-alcohol formula is very effective against bacteria, viruses and yeasts. It combines normal hand cleaning with disinfection on one procedure. In addition, no perfume or dyes are incorporated and allergic reactions are thus reduced.
Look: white foam
Color: Transparent
Smell: Alchohol Smell
Certifications: Certified with EN 1499.
Caution: Use biocides safely. Always read the label and product information before use.