Water plus - total ATP, Clean-Trace™
Catalog # MMMPAQT200
Fournisseur: 3M Food Safety
Spécifications
- Désignation:Clean-Trace™ water plus total ATP swabs
- Seq:16
- Cdt:100
Spécifications
À propos de cet article
3M™ Clean-Trace™ Water Plus – Total ATP is designed to rapidly assess equipment sanitation processes and the efficiency of Clean In Place (CIP) procedures by the improved detection of organic contamination from microbial or product residues.
- Flexible storage options: 12 months shelf life refrigerated (2 to 8 °C) or 2 months shelf life at room temperature (21 to 25 °C)
- Rapid results allowing for corrective action to be taken immediately
- Simple to use – ideal for busy production environments
- Ability to track and trend results using 3M™ Clean-Trace™ Data Trending software
The test gives an objective indication of cleanliness in water samples in seconds. Designedfor use with the 3M™ Clean-Trace™ Luminometer, the new AQT200 formulation nowprovides the ability to gain more consistent and reliable results providing greater confidencein your hygiene monitoring procedures.The new and improved formulation also provides the flexibility to store AQT200 at your ownconvenience - room temperature for up to 2 months or refrigerated for up to 12 months.