Wipe rolls, WYPALL* L20 EXTRA+
Catalog # KIMB7334
Supplier: KIMBERLY CLARK
Specifications
- Emballage:140 sheets/roll
- Couleur:bleu
- Désignation:Small roll wipes, 2-ply
- L×l:380×240 mm
- Cdt:24 Roll
Specifications
About this item
WypAll® wiping paper products are designed for the specific environments where they will be used, to deliver on your wiping task, every time.
- Versatile and small
- Easy to handle
- Perfect for tool and parts clean-up in process and maintenance environments
The embossing technology delivers a great balance of performance and value. These rolls are compatible with our WypAll® small roll dispenser (KIMB7041).