i7100 Vial Label Applicator
N° de catalogue BRDY151156
Fournisseur: Brady
Spécifications
- Désignation:i7100 vial label applicator
- Environmentally Preferable:
- Gamme de température:5 - 40°C
- Dimensions:120,65W×220,98L×76,2D mm
- Cdt:1
- Pour:BradyPrinter i7100
Spécifications
À propos de cet article
Automate vial tube identification with the Vial Label Applicator accessory that prints and applies labels on vial tubes in a few seconds.
Works only with the 300 and 600 dpi Peel models of the BradyPrinter i7100 Industrial Label Printers.
Livraison: The accessories for the wraptor printer applicator includes cleaning kits, memory cards, additional hardware, shippping materials, and stylus.