Sling polyester webbing, Protecta®, 3M™
Sling polyester webbing, 0,6 kg
N° de catalogue MMMAAM450/80
Fournisseur:   3M
Sling polyester webbing, Protecta®, 3M™
N° de catalogue MMMAAM450/80
Fournisseur:  3M
Sling polyester webbing, Protecta®, 3M™
Sling polyester webbing, Protecta®, 3M™
Sling polyester webbing, 0,6 kg
Prix
16,20 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
The Protecta® Nylon Webbing Sling Anchor features four safety seams, and measures 80 cm in length.
  • Sturdy, durable design
The Protecta® Nylon Webbing anchor straps are built from top quality materials guaranteeing high strength and durability to stand up to the toughest environments. Anchorage connectors are a critical component in a fall protection system as they securely attach the users system to the anchorage. The choice of anchor depends on the type of work being done. Anchorages are selected by what they connect to (general/all purpose, steel, concrete, roof, or specialty), whether they are fixed or mobile, and whether they are permanent or portable.
Spécifications
  • Désignation:
    Sling polyester webbing
  • Poids:
    0,6 kg