Portoir à cônes Imhoff en acrylique, Nalgene®
532-0100
- Désignation:Portoir pour 3 cônes Imhoff
- Environmentally Preferable:Low Manufacturing Impact
- Certification environnementale:NAR Certificate of Retirement
- Cdt:1
Securely hold three Imhoff cones with the durable and transparent Thermo Scientific™ Nalgene™ Acrylic Imhoff Cone Rack, featuring a wide base for stability on the lab bench.
- Transparent; easily see level of contents
- Can also be used to hold 500 and 1000 ml Nalgene separatory funnels