Welding safety spectacle, PW68
N° de catalogue PRTWPW68BGR
Fournisseur: Portwest
Spécifications
- Couleur:nuance 5
- Désignation:Welding safety spectacle, bottle green
- Cdt:1
Spécifications
À propos de cet article
Multipurpose Shade 5 lens with lateral protection and adjustable arms for customised comfort.
- 99% UV protection
- Anti-scratch coating for added durability
- Free spec cord
- Adjustable length for a secure and comfortable fit
- Excellent side protection
- Ultra lightweight for long uses
- Individually packed for vending machines
- Retail bag which aids presentation for retail sales