Endurance visor replacement, PW56
Replacement visor, clear, PW56, PC
N° de catalogue PRTWPW56CLR
Fournisseur: Portwest
Aller à :
Product Details & Documents
Impact resistant polycarbonate replacement visor is suitable for endurance helmets.
- Anti-scratch coating for added durability
- Anti-fog coating for additional comfort to the wearer
Suitable for PW54, PW55 and PS55 helmets.
Certifications: Standard: EN 166 1 FT.
Packaging: Individually packed for vending machines.
Spécifications
- Désignation:Replacement visor, clear
- Matière:PC
- Modèle:PW56