Fall protection harnesses, 3 point comfort plus
Harness, comfort plus, 2,4 kg, 3-point
N° de catalogue PRTWFP18RER
Fournisseur: Portwest
Aller à :
Product Details & Documents
Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collision and restricting users from fall hazard areas.
- Extra padding
- Attachment hoops
- Side D-rings
- Chest loops
- Made from nylon webbing
Features include two seating loops and two lower back rings. Fully adjustable with shoulder, back and seat padding for extra comfort.
Certifications: EN 361, EN 358
Spécifications
- Points d'attache:3-point
- Désignation:Harness, comfort plus
- Poids:2,4 kg