Fall protection harnesses, 2 point with chest loops
Harness with chest loop, 1,0 kg, 2-point
N° de catalogue PRTWFP12RER
Fournisseur: Portwest
Fall protection harnesses, 2 point with chest loops
N° de catalogue PRTWFP12RER
Fournisseur: Portwest
Aller à :
Product Details & Documents
Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collision and restricting users from fall hazard areas.
- Made from polyester webbing, alloy steel, heat treated
- Light weight
- Dorsal D-ring
- Chest and leg adjustment points
Features two chest loops and an adjustable quick release chest strap.
Certifications: EN 361
Spécifications
- Points d'attache:2-point
- Désignation:Harness with chest loop
- Poids:1,0 kg