Levo spectacle, PW11
Catalog # PRTWPW11CLR
Supplier: Portwest
Specifications
- Type:UV protection
- Couleur:incolore
- Oculaires:Clear PC
- Cdt:1
Specifications
About this item
Levo spectacle's innovative concept offers the comfort of a safety spectacle with the fit and seal of a goggle. The back foam gives added brow and cheek protection. The arms can be interchanged with the adjustable head-strap for more fitted wear. Excellent ventilation through indirect vents in the back foam, which aids in preventing misting and fogging.
- 99% UV protection
- Anti-scratch coating for added durability
- Anti-fog coating for additional comfort to the wearer
- Dual use with classic arms or with wide elastic band
- Detachable foam offers additional protection against shocks and dust
- Free spec cord
Materials: Polycarbonate
Range: PPE
Certifications: CE certified, EN 166 1 FT, EN 170 2C-1.2, ANSI/ISEA Z87
Packaging: Individually packed for vending machines