
4-Amino-2,3-diméthyl-1-phényl-3-pyrazoline-5-one
Catalog # MOLE43057307-500G
Supplier: Molekula
CAS Number:
Specifications
- Type de conditionnement:Glass bottle for solids
- Cdt:500 g
- Seq:3645
Specifications
About this item
:
Merck Index:13,00596
Melting Pt:107…109 °C
:203.24 g/mol
EINECS:201-452-3
:GHS07
Storage Temperature:Température ambiante
CAS Number:83-07-8
Formula:C₁₁H₁₃N₃O
MDL Number:MFCD00003145