Order Entry
Belgium
ContactUsLinkComponent
2-(4-Morpholinyl)-8-phényl-4H-chromén-4-one ≥98.0% (par HPLC)
2-(4-Morpholinyl)-8-phényl-4H-chromén-4-one ≥98.0% (par HPLC)
  TCIAM2410-25MG
 :  TCI
 :  
undefined
2-(4-Morpholinyl)-8-phényl-4H-chromén-4-one ≥98.0% (par HPLC)
  TCIAM2410-25MG
 :  TCI
 :  

 

  • Cdt:
    25 mg
  • Seq:
    2315
  • Environmentally Preferable:

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Elemental analysis (Nitrogen):
    4.40 to 4.70 %
  • Melting point:
    182.0 to 186.0 deg-C

 

 

 : LY294002, LY 294002, 2-Morpholino-8-phenyl-4H-chromen-4-one, 2-Morpholino-8-phenyl-4-oxo-4H-1-benzopyran
 :
 :307.35 g/mol
 :Congélateur
 :154447-36-6
 :C₁₉H₁₇NO₃
 :MFCD00270881