Order Entry
Belgium
ContactUsLinkComponent
Vert clair SF indicateur
Vert clair SF indicateur
Catalog # TCIAL0141-1G
Supplier:  TCI
CAS Number:  
Vert clair SF indicateur
Catalog # TCIAL0141-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Cdt:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Red to Dark red to Dark purple powder to crystal
  • Lambda max.:
    630.0 to 634.0 nm (H2O)
  • Absorbance (E1%1cm):
    min. 700 (H2O, 630.0 to 634.0 nm)

Specification Test Results

About this item

 :
 :792.86 g/mol
Merck Index:14,05485
EINECS:225-906-5
CAS Number:5141-20-8
Formula:C₃₇H₃₄N₂Na₂O₉S₃
MDL Number:MFCD00012121