Irinotecan hydrochloride trihydrate ≥98.0% (par HPLC, analyse par titration)
Catalog # TCIAI0714-100MG
Supplier: TCI
CAS Number:
Specifications
- Cdt:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Specific rotation [a]20/D:+22.0 to +26.0 deg (C=1, methanol)
- Water:7.0 to 10.0 %
Specification Test Results
About this item
:
:677.19 g/mol (623.15 anhy)
Merck Index:14,05091
Boiling Pt:257°C
:GHS07
:GHS08
Storage Temperature:Réfrigérateur
CAS Number:136572-09-3
:MFCD01862255
Formula:C₃₃H₃₈N₄O₆·HCl·3H₂O
MDL Number:MFCD01765731