
Fasudil hydrochloride ≥98.0% (par HPLC)
TCIAF0839-100MG
: TCI
:
- Cdt:100 mg
- Seq:2440
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):12.0 to 13.5 %
:
:327.83 g/mol
:14,03942
:GHS07
:105628-07-7
:C₁₄H₁₇N₃O₂S
:MFCD00943198