
4-[(E)-(4-Isothiocyanatophényl)diazényl]-N,N-diméthylaniline ≥95.0% (par analyse titrimétrique)
Catalog # TCIAD1653-100MG
Supplier: TCI
CAS Number:
Specifications
- Cdt:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Orange to Brown powder to crystal
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:169.0 to 172.0 deg-C
Specification Test Results
About this item
:
:282.37 g/mol
EINECS:231-521-3
:GHS07
CAS Number:7612-98-8
Formula:C₁₅H₁₄N₄S
MDL Number:MFCD00004812