Drop-out mix synthetic minus adenine, histidine, leucine, tryptophan, uracil without yeast nitrogen base
N° de catalogue USBID9540-05
Fournisseur: US Biological
Numéro CAS:
Spécifications
- Cdt:100 g
- Désignation:D9540 Drop-out Mix Synthetic Minus Histidine, Leucine, Tryptophan, Uracil without Yeast Nitrogen Base (DO, Dropout, Powder)
Spécifications
À propos de cet article
Drop-out mixes are used as a supplement to Drop-out base for making synthetic, complete media formulations in accordance with methods in yeast genetics.
- Convenient powders
- Useful for one-hybrid and two-hybrid systems
All lots are quality control tested using the appropriate yeast strains and transformants.