Malate standard for IC, TraceCERT®, Supelco®
Malate in water, 1000 mg/l, 100 mL
N° de catalogue 06994-100ML
Fournisseur: Merck
Malate standard for IC, TraceCERT®, Supelco®
N° de catalogue 06994-100ML
Fournisseur: Merck
Numéro CAS:
Aller à :
Product Details & Documents
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Spécifications
Spécifications
- Concentration:1000 mg/l
- Cdt:100 mL
- Désignation:Malate in water