Order Entry
Belgium
ContactUsLinkComponent
VWR®, Jar, cylindrical, screw neck
VWR®, Jar, cylindrical, screw neck
Catalog # LAPH45092154
Supplier:  LA PHA PACK
VWR®, Jar, cylindrical, screw neck
Catalog # LAPH45092154
Supplier:  LA PHA PACK

Specifications

  • Ø ext.:
    48,5 mm
  • Capacité:
    100 ml
  • Hauteur:
    85,5 mm
  • Cdt:
    72

Specifications

About this item

Made of clear glass.