Supelco® Organic Reference Standard, BDE No 126 Solution
BDE No 126 solution, Analytical standard, 1 mL
  33682-1ML
 :   Merck
Supelco® Organic Reference Standard, BDE No 126 Solution
  33682-1ML
 :  Merck
 :  
Supelco® Organic Reference Standard, BDE No 126 Solution
Supelco® Organic Reference Standard, BDE No 126 Solution
BDE No 126 solution, Analytical standard, 1 mL
 :  
457,00 €
1 * 1 mL
  • GHS signal: GHS02, GHS08, GHS07, GHS09
 : P210 - P273 - P301 + P310 + P331 - P302 + P352
  • Cdt:
    1 mL
  • Normes:
    Analytical standard
  • Désignation:
    BDE No 126 solution
  • Cat. No.:
    33682-1ML
  • N ° CAS.:
    366791-32-4
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₂H₅Br₅O
  • Conditions de stockage:
    2...8 °C
  • Description de la classe de danger:
    H225 - H304 - H315 - H336 - H410
  • Point d'éclair:
    –12 °C
  • Conditions de transport:
    STD
  • Synonyme(s):
    3,3′,4,4′,5-Pentabromodiphenyl ether solution; 3,3′,4,4′,5-PentaBDE; PBDE 126
  • MDL No.:
    MFCD09265278
  • Masse moléculaire:
    564,69
  • Température de stockage:
    2...8 °C