Test strips, cyanuric acid, swimming pool test
N° de catalogue MANA90710
Fournisseur: MACHEREY-NAGEL
Numéro CAS:
Spécifications
- Cdt:1
- Désignation:Cyanuric acid test strips
- Seq:03
- Tests par set:25
- Plage de mesure:0 - 300 mg/l
Spécifications
À propos de cet article
These test strips allow the quick and easy determination of the water quality in swimming pools. The test provides valuable results about the Cyanuric acid content.
- Reliable result is obtained within 30 seconds.
- Just dip, wait and read against multi-stage color scale
- Always ready to use
- No accessories necessary for evaluation
- Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
The test strip is dipped into the pool and moved back and forth 5 times.
Monitoring of the water quality allows adjusting the dosage of water conditioners in good time. The pool remains nice and clean.