Safety spectacles, uvex x-fit
N° de catalogue UVEX9199.085
Fournisseur: UVEX
Spécifications
- Type:Anti-UV (UV 400), anti-scratch, anti-fog
- Couleur:gris
- Oculaires:PC clear
- Cdt:1
Spécifications
À propos de cet article
The model x-fit are classic safety spectacles with good coverage of the eyes and a high level of wearer acceptance.
- Supravision coating technology
- Metal-free
- Translucent side arms
Weight: 23 g
Certifications: EN 166; EN 170