Earmuffs, K2
Earmuffs, uvex K2, jaune/noir, 32 dB
N° de catalogue UVEX2600.002
Fournisseur: UVEX
Aller à :
Product Details & Documents
These K2 protective earmuffs offer an impressive combination of high levels of protection combined with great design of soft surfaces. These earmuffs are easy to mould and comfortable to wear due to their low weight.
- Easy length adjustment
- Easily adjustable for a perfect fit
- Padded headband for optimal comfort
- Extra-soft memory foam ear cushions
- Designed to be lightweight with a small capsule
SNR: 32 dB
Certifications: EN 352-1
Spécifications
- Hauteur du plateau:32 dB
- Couleur:jaune/noir
- Désignation:Earmuffs, uvex K2