Imaging reference target for myECL™ imager
Catalog # PIER62267
Supplier: Thermo Fisher Scientific
Specifications
- Désignation:Imaging reference target for myECL™ imager
- Cdt:1
Specifications
About this item
Plastic imaging target with printed design; 7×6,5 inches (178×165 mm). This product is a replacement for the myECL™ Imager accessory used for focusing in all imaging modes.