SBW glass stoppers, standard ground joint, octagonal
Catalog # 217-9125
Supplier: DWK Life Sciences
Specifications
- RN:14/23
- Type:Verre transparent
- Seq:12
- Cdt:1
Specifications
About this item
These stoppers are made from SBW glass and have an octagonal head for good grip.
- Solid form
- Available amber or clear
Certifications: DIN 12252