VISOCOLOR® Colorimetric Tests, alpha Residual Hardness
MANA935080
:
- Méthode chimique:Mixed indicator
- Cdt:1 KIT
- Désignation:VISOCOLOR® alpha Residual Hardness
- Dimensions:150×93×35 mm (Including packaging)
- Plateforme:Colorimetric test kits
- Température de stockage:15...25 °C
- Durée de conservation (mois):12
- Poids:47,7 g (Including packaging)
- Pour appareil:Visual
- Tests par set:200
- Plage de mesure:0,00 • 0,05 • 0,10 • 0,19 • 0,38 °e
Colorimetric test kit for the simplest determination of residual hardness.
- Combines simple implementation with high measurement reliability
VISOCOLOR alpha residual hardness is the simplest colorimetric test kit for residual hardness in the VISOCOLOR range.
Sea water analysis: No
Hazardous material: Yes
:
Reagents for 200 tests and colour chart in a blister.