Aprons for food industry, Nantou 8195
: SIOEN
The Nantou is a low-noise and durable apron made from 100% polyamide 6.6 knitting with PU coating.
- Waterproof, flexible and elastic Flexothane® Kleen fabric
- Neck strap with buckles
- Two cords on the waist and extra stomach reinforcement
- Washable at 95 °C for optimum hygiene
Developed for the food industry.
Weight: 170 g/m²
Colour: White or blue
: EN 14605 type PB 4, EN 340:2003, (EC) 1935:2004, Food contact approved
- Catalog No:
- SIOE8195A2FK0W02
- SIOE8195A2FK0H45
- Applications des tenues:
- Alimentaire
- Alimentaire
- Zone à risque:
- Pluie (imperméable)
- Pluie (imperméable)
- Genre:
- Unisex
- Unisex
- Type de fermeture:
- Avec fermeture dans le dos
- Avec fermeture dans le dos
- Tissu:
- 100% polyamide 6,6 knitting
- 100% polyamide 6,6 knitting
- Poids:
- 170 g/m²
- 170 g/m²
Frequently Bought Together