Test strips for semi-quantitative analysis of low water hardness, AQUADUR® sensitive
N° de catalogue MANA91243
Fournisseur: MACHEREY-NAGEL
Numéro CAS:
Spécifications
- Cdt:50
- Désignation:Aquadur® sensitive test strips, box, box 6×95 mm
- Plage de mesure:< 0,5 - >18,8 mg/l CaCO₃
Spécifications
À propos de cet article
AQUADUR® sensitive test strips for semi-quantitative analysis of low water hardness, after amongst other things the softening of water.
Livraison: Supplied in one aluminium container with 50 test strips.