Cutting board
Cutting board
Cutting board, bleu, 500×400×20 mm
N° de catalogue KPXX060.012.119
Fournisseur:   KLINIPATH
Cutting board
N° de catalogue KPXX060.012.119
Fournisseur:  KLINIPATH
Cutting board
Cutting board, bleu, 500×400×20 mm
Prix
362,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
The cutting board is completely made of PE.
The top thickness is approx. 20 mm. The board includes 4 small feet of 25 mm height and 15 mm diameter.
Attention: Not suitable for thermal sterilisation.
Spécifications
  • Couleur:
    bleu
  • Désignation:
    Cutting board
  • l×p×h:
    500×400×20 mm