Acetate mix (5 components)
8260 Acetate Mix, 2000 µg/ml each in P and T methanol, 1 ml/ampoule, 1 mL
N° de catalogue REST30477
Fournisseur: Restek
Aller à :
Product Details & Documents
This contains: n-butyl acetate; ethyl acetate; isopropyl acetate; n-propyl acetate; vinyl acetate.
Spécifications
Spécifications
- Concentration:2000 µg/ml each in P and T methanol
- Cdt:1 mL
- Désignation:8260 Acetate Mix
- Info cdt:1 ml/ampoule