VOA tuning compound
1-Bromo-4-fluorobenzene (BFB), 5000 μg/ml in P and T methanol, 1 ml/ampule, 1
N° de catalogue REST30003
Fournisseur: Restek
Aller à :
Product Details & Documents
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Spécifications
Spécifications
- Concentration:5000 μg/ml in P and T methanol
- Cdt:1
- Désignation:1-Bromo-4-fluorobenzene (BFB)
- Info cdt:1 ml/ampule
- Cat. No.:REST30003
- N ° CAS.:460-00-4
- Formula:C₆H₄BrF