508 performance check mix (4 components)
508 performance check mix (4 components)
508 performance check mix, In methyl tert-butyl ether, 1 ml/ampoule, 1 mL
N° de catalogue REST32045
Fournisseur:   Restek
508 performance check mix (4 components)
N° de catalogue REST32045
Fournisseur:  Restek
Numéro CAS:  
508 performance check mix (4 components)
508 performance check mix (4 components)
508 performance check mix, In methyl tert-butyl ether, 1 ml/ampoule, 1 mL
Numéro CAS:  
Prix
119,00 €
Connexion Pour la disponibilité
1 * 1 mL
Product Details & Documents
This mix contains four components.
Components are δ-BHC (0,4 μg/ml), chlorothalonil (0,5 μg/ml), chlorpyrifos (0,02 μg/ml), DCPA methyl ester (Chlorthal-dimethyl) (0,5 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Spécifications
Spécifications
  • Concentration:
    In methyl tert-butyl ether
  • Cdt:
    1 mL
  • Désignation:
    508 performance check mix
  • Info cdt:
    1 ml/ampoule