LLL liquid detergent
LLL liquid detergent
  LANC41010070
LLL liquid detergent
  LANC41010070
LLL liquid detergent
LLL liquid detergent
370,00 €
1 * 10 L
LLL detergent can be used for an efficient and consistent washing process.
  • Phosphate-free
  • Non foaming
LLL detergent assures high performance washing by a contamination-free process with compatibility between equipment material and chemicals used.
  • Désignation:
    LLL liquid detergent