Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Scellés
Films de scellage
Cling film in dispenser box
Cling film in dispenser box
Cling film in dispenser box
KING756030
:
KING DISPOSABLES
Cling film in dispenser box
KING756030
:
KING DISPOSABLES
Largeur:
30 cm
Désignation:
Cling film in dispenser box, 9 µm thickness
Longueur:
300 m
Matière:
PVC
Cdt:
6