Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Retention time marker

Retention time marker

Retention time marker, 1 ml/ampoule, 1
N° de catalogue REST30483
Fournisseur:   Restek
Retention time marker
N° de catalogue REST30483
Fournisseur:  Restek
Numéro CAS:  
Retention time marker

Retention time marker, 1 ml/ampoule, 1

Numéro CAS:  
Prix
64,40 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

This standard contains three components, 1000 µg/ml each in P and T methanol

Components: n-hexane (C6), n-decane (C10) and n-dodecane (C12).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

Spécifications

Spécifications

  • Cdt:
    1
  • Désignation:
    Retention time marker
  • Info cdt:
    1 ml/ampoule

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.