Huile à immersion, BioChemica
Catalog # A3494.0500
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Cdt:500 mL
- Désignation:Immersion oil
Specifications
About this item
Biochemical grade immersion oil for microscopy.
- Free of PCBs
- Stored at room temperature