Silver capsule for oxygen analysis kit
N° de catalogue PERCN2411364
Fournisseur: PerkinElmer
Spécifications
- Désignation:Silver capsules for oxygen analysis kit
- Environmentally Preferable:
- Cdt:96
Spécifications
À propos de cet article
Silver capsule used for analysing organic substances. For oxygen content either silver or standard tin capsules may be used for analysing organic substances for oxygen content.