2-Naphthyl boronic acid
Glass bottle for solids, 1 g
Catalog # MOLEM51174107
Marketsource
Supplier: Molekula
2-Naphthyl boronic acid
Catalog # MOLEM51174107
Supplier: Molekula
Marketsource
CAS Number: 32316-92-0
Jump to:
Product Details & Documents
Synonyms: 2-Naphthaleneboronic acid, Naphthalene-2-boronic acid, 2-Naphthyl boric acid
- CAS Number:32316-92-0
- Formula:C₁₀H₉BO₂
- MDL Number:MFCD00236051
- Molecular Weight:171.99 g/mol
Specifications
Specifications
- Type de conditionnement:Glass bottle for solids
- Cdt:1 g



