Potassium dihydrogénophosphate
Catalog # MOLE42899676-1KG
Supplier: Molekula
CAS Number:
Specifications
- Type de conditionnement:Glass bottle for solids
- Cdt:1 kg
- Seq:4024
Specifications
About this item
:
:136.09 g/mol
Merck Index:13,07744
Boiling Pt:∼100 °C (1013 hPa)
Melting Pt:253 °C
Density:2.3 g/cm³ (20 °C)
Storage Temperature:Température ambiante
EINECS:231-913-4
REACH:01-2119490224-41
CAS Number:7778-77-0
Formula:KH₂PO₄
MDL Number:MFCD00011401