2,2',4-Trichlorobiphényle
Glass bottle for solids, 5 mg
N° de catalogue PROOCIL-PCB-17-C
Fournisseur: LGC Standards PROMOCHEM
2,2',4-Trichlorobiphényle
N° de catalogue PROOCIL-PCB-17-C
Fournisseur: LGC Standards PROMOCHEM
Numéro CAS: 37680-66-3
Aller à :
Product Details & Documents
- Numéro CAS:37680-66-3
- Formule:C₁₂H₇Cl₃
- Poids moléculaire:257.55 g/mol
Spécifications
Spécifications
- Type de conditionnement:Glass bottle for solids
- Cdt:5 mg
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



