Pyrogallol
Glass bottle for solids, 100 g
Catalog # ROTH3963.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene
- CAS Number:87-66-1
- Formula:C₆H₆O₃
- MDL Number:MFCD00002192
- Molecular Weight:126.11 g/mol
- Merck Index:13,08090
- Boiling Pt:309 °C (1013 hPa)
- Melting Pt:130…132 °C
- Density:1,453 g/cm³ (20 °C)
- Storage Temperature:Température ambiante
- EINECS:201-762-9
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Type de conditionnement:Glass bottle for solids
- Cdt:100 g