Order Entry
Belgium
ContactUsLinkComponent
Acide (3-bromophényl)acétique
Acide (3-bromophényl)acétique
Catalog # MOLE34023257-5G
Supplier:  Molekula
CAS Number:  
Acide (3-bromophényl)acétique
Catalog # MOLE34023257-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Type de conditionnement:
    Glass bottle for solids
  • Cdt:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :215.05 g/mol
Boiling Pt:308 °C (1013 hPa)
Melting Pt:102…103 °C
EINECS:217-522-1
CAS Number:1878-67-7
Formula:(C₆H₄Br)CH₂COOH
MDL Number:MFCD00004330