Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Agitateurs
Agitateurs rotatifs
Platform rocker, tapped density tester
Platform rocker, tapped density tester
Platform rocker, tapped density tester
VARI52-1080
:
VARIAN
Platform rocker, tapped density tester
VARI52-1080
:
VARIAN
Désignation:
Platform, 250 ml, 3 mm drop, USP or ASTM
Cdt:
1