ContactUsLinkComponent
Bio-7-dATP
Bio-7-dATP
Catalog # ENZOENZ42819
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
undefined
Bio-7-dATP
Catalog # ENZOENZ42819
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Specifications
  • Cdt:
    50 Nmo
  • Hôte:
  • Désignation:
    Bio-7-dATP
  • Seq:
    02
  • Environmentally Preferable:
Specifications
About this item
Bio-7-dATP (Biotin-7-2’-deoxyadenosine-5’-triphosphate) can replace dATP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase.
Bio-7-dATP can be used to produce biotinylated DNA probes in a variety of labelling reactions including nick translation, random prime labelling, cDNA labelling and 3’-end labelling. The resulting biotin-labelled probe can be used in a variety of hybridisation applications incuding Southern blots, Northern blots, or dot blots. The probes can also be used for in situ hybridisation procedures on fixed cells and tissues. Probes have been shown to hybridize to homologous nucleic acid at the same rate and to the same extent as non-biotinylated probes. They can be detected by their interaction with biotin-binding proteins, such as avidin, streptavidin or antibodies coupled to fluorescent dyes or colour-producing enzymes.