Stop solution 2
Stop solution 2
Stop solution 2, 50 mL
N° de catalogue ENZOADI801804
Fournisseur:   Enzo Life Sciences
Stop solution 2
N° de catalogue ENZOADI801804
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Stop solution 2
Stop solution 2, 50 mL
Numéro CAS:  
Prix
94,60 €
Connexion Pour la disponibilité
1 * 50 mL
Product Details & Documents
Acid stop solution (1 N HCl) intended for use as an assay stop reagent for colorimetric HRP/TMB reactions.
Each bottle is sufficient for use with 5×96 wells microtiter plates.
Informations de commande: Additional assay development reagents are available as companion products to IMMUNOSET® products.
Spécifications
Spécifications
  • Cdt:
    50 mL
  • Désignation:
    Stop solution 2
  • Cat. No.:
    ENZOADI801804
  • Conditions de stockage:
    Ambient