Potassium hydroxyde 0.1 mol/l (0.1 N) en solution aqueuse
Supplier: PROLABO CMP
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Storage Temperature:Température ambiante
EINECS:215-181-3
UN:1814
ADR:8,II
:GHS05
Frequently Bought Together