Neutralising reagent
Neutralising reagent, 5 mL
N° de catalogue ENZOADI801475
Fournisseur: Enzo Life Sciences
Aller à :
Product Details & Documents
Neutralising buffer for low pH solutions and for use with cyclic nucleotide and isoprostane kits.
Spécifications
Spécifications
- Cdt:5 mL
- Désignation:Neutralising reagent