Dichlorométhane ≥99.5% stabilisé ACS
About this item
Purity: 99.5+%
CAS No.: 75-09-2
Molecular Formula: CH2Cl2
Formula Weight: 84.93
Physical Form: Liquid
Density: 1.325
Refractive Index: 1.4244
Appearance: Colourless
Odour: Sweet
Melting Point: -95°
Boiling Point: 39-40°
Beilstein Reference: 1730800
Merck Reference: 14,6063
UN No.: 1593
MDL No.: MFCD00000881
- CAS Number:75-09-2
- Formula:CH₂Cl₂
- MDL Number:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck Index:14,6063
- Boiling Pt:39…40 °C
- Melting Pt:–95 °C
- Density:1,325
- Storage Temperature:Température ambiante
- EINECS:200-838-9
- ADR:6.1,III
- :Stabilisé avec de l'Amylène
- UN:1593
3 Options Available Below
Product Details & Documents
Purity: 99.5+%
CAS No.: 75-09-2
Molecular Formula: CH2Cl2
Formula Weight: 84.93
Physical Form: Liquid
Density: 1.325
Refractive Index: 1.4244
Appearance: Colourless
Odour: Sweet
Melting Point: -95°
Boiling Point: 39-40°
Beilstein Reference: 1730800
Merck Reference: 14,6063
UN No.: 1593
MDL No.: MFCD00000881
- CAS Number:75-09-2
- Formula:CH₂Cl₂
- MDL Number:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck Index:14,6063
- Boiling Pt:39…40 °C
- Melting Pt:–95 °C
- Density:1,325
- Storage Temperature:Température ambiante
- EINECS:200-838-9
- ADR:6.1,III
- :Stabilisé avec de l'Amylène
- UN:1593
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information